Stampa…

5-Chloro-2-(methylthio)benzothiazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C2111-5G C2111-1G
TCIAC2111-5GEA 174 EUR
TCIAC2111-5G TCIAC2111-1G
5-Chloro-2-(methylthio)benzothiazole ≥98.0% (by GC)
5-Chloro-2-(methylthio)benzothiazole
Formula: C₈H₆ClNS₂
CAS Number: 3507-41-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR