Stampa…

4'-Chloropivaloanilide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-(4-Chlorophenyl)pivalamide, 4-Chloro-N-pivaloylaniline, N-(4-Chlorophenyl)-2,2-dimethylpropionamide

C2099-25G C2099-250G
TCIAC2099-25GEA 52 EUR
TCIAC2099-25G TCIAC2099-250G
4'-Chloropivaloanilide ≥98.0% (by GC)
4'-Chloropivaloanilide
Formula: C₁₁H₁₄ClNO
Peso molecolare: 211,69 g/mol
MDL Number: MFCD01571825
CAS Number: 65854-91-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR