Stampa…

5-(Carbobenzoxyamino)valeric acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 5-(Benzyloxycarbonylamino)valeric acid, 5-(Carbobenzoxyamino)pentanoic Acid, N-Cbz-5-aminovaleric Acid, 5-(((Benzyloxy)carbonyl)amino)pentanoic acid, N-Cbz-5-aminopentanoic Acid

C2067-25G C2067-5G
TCIAC2067-25GEA 725 EUR
TCIAC2067-25G TCIAC2067-5G
5-(Carbobenzoxyamino)valeric acid ≥98.0% (by titrimetric analysis)
5-(Carbobenzoxyamino)valeric acid
Formula: C₁₃H₁₇NO₄
Peso molecolare: 251,28 g/mol
MDL Number: MFCD00188458
CAS Number: 23135-50-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 103.0 to 107.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR