Stampa…

Cyclopropanecarboxamide ≥98.0% (by GC)

Fornitore: TCI EUROPE

C2010-25G C2010-250G
TCIAC2010-25GEA 50 EUR
TCIAC2010-25G TCIAC2010-250G
Cyclopropanecarboxamide ≥98.0% (by GC)
Cyclopropanecarboxamide
Formula: C₄H₇NO
Peso molecolare: 85,11 g/mol
MDL Number: MFCD00013729
CAS Number: 6228-73-5
EINECS: 228-332-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR