2-Cyanoethyl acrylate ≥95.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Acrylic Acid 2-Cyanoethyl Ester, 3-Acryloyloxypropionitrile
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
C1974-25G
C1974-5G
TCIAC1974-25GEA
396
EUR
TCIAC1974-25G
TCIAC1974-5G
2-Cyanoethyl acrylate ≥95.0% (by GC) stabilizzato
2-Cyanoethyl acrylate
Formula:
C₆H₇NO₂ Peso molecolare: 125,13 g/mol Punto di ebollizione: 108°C Flash Pt: 124°C |
MDL Number:
MFCD00013823 CAS Number: 106-71-8 EINECS: 203-426-7 UN: 3276 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...