2'-(2-Chloroanilino)-6'-(dibutylamino)fluoran ≥97.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
Synonyms:
2'-(2-Chlorophenylamino)-6'-(dibutylamino)fluoran
C1946-25G
C1946-500G
TCIAC1946-25GEA
38
EUR
TCIAC1946-25G
TCIAC1946-500G
2'-(2-Chloroanilino)-6'-(dibutylamino)fluoran ≥97.0% (by HPLC, total nitrogen)
2'-(2-Chloroanilino)-6'-(dibutylamino)fluoran
Formula:
C₃₄H₃₃ClN₂O₃ Peso molecolare: 553,10 g/mol |
CAS Number:
82137-81-3 EINECS: 279-906-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 184.0 to 188.0 deg-C |
Solubility in hot Toluene | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...