Stampa…

6-Chloropicolinic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Chloropyridine-2-carboxylic acid, 2-Chloropyridine-6-carboxylic acid

C1896-1G C1896-5G
TCIAC1896-1GEA 17 EUR
TCIAC1896-1G TCIAC1896-5G
6-Chloropicolinic acid ≥98.0% (by GC, titration analysis)
6-Chloropicolinic acid
Formula: C₆H₄ClNO₂
Peso molecolare: 157,56 g/mol
MDL Number: MFCD00155390
CAS Number: 4684-94-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 186.0 to 190.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR