Stampa…

3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-Chloro-2,2'-iminodibenzyl

C1866-25G C1866-5G
TCIAC1866-25GEA 212 EUR
TCIAC1866-25G TCIAC1866-5G
3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine ≥98.0% (by GC)
3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine
Formula: C₁₄H₁₂ClN
Peso molecolare: 229,71 g/mol
MDL Number: MFCD01632171
CAS Number: 32943-25-2
EINECS: 251-301-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b97c94eab1c9925032ac333791215e09de64116&vid=9b97c94eab1c9925032ac333791215e09de64116&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR