3-Chloro-2,4(3H,5H)-furandione ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Chlorotetronic Acid, 3-Chlorofuran-2,4(3H,5H)-dione, 3-Chlorotetrahydrofuran-2,4-dione, 3-Chloro-4-hydroxy-2(5H)-furanone
C1810-25G
C1810-5G
TCIAC1810-25GEA
261
EUR
TCIAC1810-25G
TCIAC1810-5G
3-Chloro-2,4(3H,5H)-furandione ≥98.0% (by GC, titration analysis)
3-Chloro-2,4(3H,5H)-furandione
Formula:
C₄H₃ClO₃ Peso molecolare: 134,52 g/mol |
MDL Number:
MFCD00134273 CAS Number: 4971-55-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...