N-Chlorosaccharin ≥96.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₇H₄ClNO₃S Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00274277 CAS Number: 14070-51-0 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (Iodometric Titration) | min. 96.0 % |
Melting point | 144.0 to 149.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...