Benzyl-4-nitrophenyl carbonate ≥96.0% (by GC)
Fornitore: TCI
Formula:
C₁₄H₁₁NO₅ |
MDL Number:
MFCD00007323 CAS Number: 13795-24-9 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 75.0 to 79.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...