N-(tert-Butoxycarbonyloxy)phthalimide ≥95.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
tert-Butyl (1,3-dioxoisoindolin-2-yl) carbonate, Carbonic Acid tert-Butyl Phthalimido Ester, tert-Butyl Phthalimido Carbonate
C1573-5G
C1573-1G
TCIAC1573-5GEA
116
EUR
TCIAC1573-5G
TCIAC1573-1G
N-(tert-Butoxycarbonyloxy)phthalimide ≥95.0% (by HPLC)
N-(tert-Butoxycarbonyloxy)phthalimide
Formula:
C₁₃H₁₃NO₅ Peso molecolare: 263,25 g/mol |
MDL Number:
MFCD00005889 CAS Number: 15263-20-4 EINECS: 239-308-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...