Stampa…

3-Chloro-2,2-dimethyl-1-propanol ≥98.0% (by GC)

Fornitore: TCI EUROPE

C1538-25G C1538-500G
TCIAC1538-25GEA 37 EUR
TCIAC1538-25G TCIAC1538-500G
3-Chloro-2,2-dimethyl-1-propanol ≥98.0% (by GC)
3-Chloro-2,2-dimethyl-1-propanol
Formula: C₅H₁₁ClO
Peso molecolare: 122,59 g/mol
MDL Number: MFCD00004684
CAS Number: 13401-56-4
EINECS: 236-488-9

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %
Freezing point 32.0 to 35.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR