Croconic acid ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4,5-Dihydroxy-4-cyclopentene-1,2,3-trione, 4,5-Dihydroxycyclopent-4-ene-1,2,3-trione
C1483-1G
C1483-5G
TCIAC1483-1GEA
129
EUR
TCIAC1483-1G
TCIAC1483-5G
Croconic acid ≥98.0% (by HPLC)
Croconic acid
Formula:
C₅H₂O₅ Peso molecolare: 142,07 g/mol Punto di fusione: 114°C |
MDL Number:
MFCD00181389 CAS Number: 488-86-8 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...