Stampa…

4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine-1-oxyl ≥96.0% (by HPLC) free radical

Fornitore: TCI
Attenzione

Synonyms: 4-(2-Chloroacetamido)-TEMPO

C1432-1G C1432-100MG
TCIAC1432-1GEA 151 EUR
TCIAC1432-1G TCIAC1432-100MG
4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine-1-oxyl ≥96.0% (by HPLC) free radical
4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine-1-oxyl
Formula: C₁₁H₂₀ClN₂O₂
Peso molecolare: 247.74 g/mol
MDL Number: MFCD00041858
CAS Number: 36775-23-2

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 96.0 area%
Melting point 117.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR