Stampa…

4-Carboxy-2,2,6,6-tetramethylpiperidine-1-oxyl ≥97.0% (by GC, titration analysis) free radical

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Carboxy-TEMPO, 4-Carboxy-2,2,6,6-tetramethylpiperidine1-oxyl

C1428-1G C1428-100MG
TCIAC1428-1GEA 291 EUR
TCIAC1428-1G TCIAC1428-100MG
4-Carboxy-2,2,6,6-tetramethylpiperidine-1-oxyl ≥97.0% (by GC, titration analysis) free radical
4-Carboxy-2,2,6,6-tetramethylpiperidine-1-oxyl
Formula: C₁₀H₁₈NO₃
Peso molecolare: 200,26 g/mol
MDL Number: MFCD00134569
CAS Number: 37149-18-1

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR