Stampa…

1,2,3,4,5,6-Cyclohexanehexacarboxylic acid ≥97.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: Cyclohexane-1,2,3,4,5,6-hexacarboxylic acid, 1,2,3,4,5,6-Cyclohexanehexacarboxylic acid monohydrate

C1427-25G C1427-5G
TCIAC1427-25GEA 328 EUR
TCIAC1427-25G TCIAC1427-5G
1,2,3,4,5,6-Cyclohexanehexacarboxylic acid ≥97.0% (by titrimetric analysis)
1,2,3,4,5,6-Cyclohexanehexacarboxylic acid
Formula: C₁₂H₁₂O₁₂
Peso molecolare: 348.22 g/mol
MDL Number: MFCD00081090
CAS Number: 2216-84-4
EINECS: 218-699-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a87018525826666fb24de554127c4de314bccaa&vid=9a87018525826666fb24de554127c4de314bccaa&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (Neutralization titration) min. 97.0% (calcd.on anh.substance)
Solubility in hot Water almost transparency
Water 4.7 to 5.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR