Stampa…

(Chloromethyl)triethoxysilane ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C1402-10G C1402-25G
TCIAC1402-10GEA 16 EUR
TCIAC1402-10G TCIAC1402-25G
(Chloromethyl)triethoxysilane ≥95.0% (by GC)
(Chloromethyl)triethoxysilane
Formula: C₇H₁₇ClO₃Si
Peso molecolare: 212,75 g/mol
Punto di ebollizione: 180°C
Flash Pt: 47°C
MDL Number: MFCD00039354
CAS Number: 15267-95-5
EINECS: 239-311-3
UN: 1993
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR