Dichlorocyclohexylmethylsilane ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Cyclohexylmethyldichlorosilane
C1399-25G
C1399-5G
TCIAC1399-25GEA
132
EUR
TCIAC1399-25G
TCIAC1399-5G
Dichlorocyclohexylmethylsilane ≥97.0% (by GC)
Dichlorocyclohexylmethylsilane
Formula:
C₇H₁₄Cl₂Si Peso molecolare: 197,17 g/mol Punto di ebollizione: 202°C Flash Pt: 66°C |
MDL Number:
MFCD00046333 CAS Number: 5578-42-7 EINECS: 226-956-0 UN: 2987 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...