Stampa…

(S)-2-Chloro-4-methylvaleric acid ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-2-Chloro-4-methylpentanoic Acid

C1371-1G C1371-5G
TCIAC1371-1GEA 76 EUR
TCIAC1371-1G TCIAC1371-5G
(S)-2-Chloro-4-methylvaleric acid ≥97.0% (by titrimetric analysis)
(S)-2-Chloro-4-methylvaleric acid
Formula: C₆H₁₁ClO₂
Peso molecolare: 150,60 g/mol
Punto di ebollizione: 126°C
MDL Number: MFCD00044943
CAS Number: 28659-81-6

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D -32.0 to -35.0deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR