[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid ≥99.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-((4-Chloro-6-((2,3-dimethylphenyl)amino)pyrimidin-2-yl)thio)acetic acid, WY-14643, Pirinixic acid
C1323-1G
C1323-5G
TCIAC1323-1GEA
148
EUR
TCIAC1323-1G
TCIAC1323-5G
[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid ≥99.0% (by HPLC, titration analysis)
[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid
Formula:
C₁₄H₁₄ClN₃O₂S Peso molecolare: 323,80 g/mol |
MDL Number:
MFCD00191335 CAS Number: 50892-23-4 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Neutralization titration) | min. 99.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...