Stampa…

4(5)-Cyanomethylimidazole ≥99.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-(4-Imidazolyl)acetonitrile

C1318-1G C1318-5G
TCIAC1318-1GEA 64 EUR
TCIAC1318-1G TCIAC1318-5G
4(5)-Cyanomethylimidazole ≥99.0% (by GC, titration analysis)
4(5)-Cyanomethylimidazole
Formula: C₅H₅N₃
Peso molecolare: 107,12 g/mol
MDL Number: MFCD00070306
CAS Number: 18502-05-1
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 138.0 to 143.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR