Stampa…

2-Chloro-2-oxo-1,3,2-dioxaphospholane ≥95.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Ethylene glycol chlorophosphate, 2-Chloro-1,3,2-dioxaphospholane-2-oxide

C1250-25G C1250-5G
TCIAC1250-25GEA 213 EUR
TCIAC1250-25G TCIAC1250-5G
2-Chloro-2-oxo-1,3,2-dioxaphospholane ≥95.0% (by GC, titration analysis)
2-Chloro-2-oxo-1,3,2-dioxaphospholane
Formula: C₂H₄ClO₃P
Peso molecolare: 142,47 g/mol
Punto di ebollizione: 91°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00043138
CAS Number: 6609-64-9
EINECS: 229-560-6
UN: 3265
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR