Stampa…

4-Chloro-2,5-dimethoxynitrobenzene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C1244-25G C1244-250G
TCIAC1244-25GEA 51 EUR
TCIAC1244-25G TCIAC1244-250G
4-Chloro-2,5-dimethoxynitrobenzene ≥98.0% (by GC)
4-Chloro-2,5-dimethoxynitrobenzene
Formula: C₈H₈ClNO₄
Peso molecolare: 217,61 g/mol
MDL Number: MFCD00024577
CAS Number: 6940-53-0
EINECS: 230-086-7

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 140.0 to 144.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR