3-(Cyanomethyl)-2,4,5-trimethylthiophene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-(2,4,5-Trimethylthiophen-3-yl)acetonitrile
C1228-1G
C1228-5G
TCIAC1228-1GEA
130
EUR
TCIAC1228-1G
TCIAC1228-5G
3-(Cyanomethyl)-2,4,5-trimethylthiophene ≥98.0% (by GC)
3-(Cyanomethyl)-2,4,5-trimethylthiophene
Formula:
C₉H₁₁NS Peso molecolare: 165,25 g/mol Punto di ebollizione: 130°C |
MDL Number:
MFCD00143113 CAS Number: 112440-49-0 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 66.0 to 69.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...