Stampa…

Cystamine sulphate hydrate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 2,2'-Diaminodiethyl Disulphide Sulphate, 2,2'-Dithiobis(ethylamine) sulphate

C1063-25G C1063-500G
TCIAC1063-25GEA 32 EUR
TCIAC1063-25G TCIAC1063-500G
Cystamine sulphate hydrate ≥98.0% (by titrimetric analysis)
Cystamine sulphate hydrate
Formula: C₄H₁₂N₂S₂
Peso molecolare: 250,35 g/mol
MDL Number: MFCD00060254
CAS Number: 16214-16-7
EINECS: 240-340-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0% (calcd.on anh.substance)
Water max. 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR