Stampa…

Chlorocarbonylsulphenyl chloride ≥97.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

Synonyms: Chlorothioformyl Chloride, Chloroformylsulphenyl Chloride

C1043-25G C1043-5G
TCIAC1043-25GEA 147 EUR
TCIAC1043-25G TCIAC1043-5G
Chlorocarbonylsulphenyl chloride ≥97.0% (by titrimetric analysis)
Chlorocarbonylsulphenyl chloride
Formula: CCl₂OS
Punto di ebollizione: 98°C
Flash Pt: 62°C
MDL Number: MFCD00000703
CAS Number: 2757-23-5
EINECS: 220-415-2
UN: 3265
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green clear liquid
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR