Isobutyl cyanoacetate ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₇H₁₁NO₂ |
MDL Number:
MFCD00044320 CAS Number: 13361-31-4 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...