Methyl-3-cyclohexene-1-carboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 3-cyclohexenecarboxylate, 3-Cyclohexene-1-carboxylic Acid Methyl Ester, Methyl cyclohex-3-enecarboxylate
C0967-25ML
C0967-500ML
TCIAC0967-25MLEA
59
EUR
TCIAC0967-25ML
TCIAC0967-500ML
Methyl-3-cyclohexene-1-carboxylate ≥98.0% (by GC)
Methyl-3-cyclohexene-1-carboxylate
Formula:
C₈H₁₂O₂ Peso molecolare: 140,18 g/mol Punto di ebollizione: 185°C Flash Pt: 60°C |
MDL Number:
MFCD00053266 CAS Number: 6493-77-2 EINECS: 229-376-6 UN: 3272 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...