Stampa…

Ethyl-2-cyano-3,3-diphenylacrylate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Cyano-3,3-diphenylacrylic Acid Ethyl Ester, Ethyl 2-cyano-3,3-diphenylacrylate

C0962-25G C0962-500G
TCIAC0962-25GEA 17 EUR
TCIAC0962-25G TCIAC0962-500G
Ethyl-2-cyano-3,3-diphenylacrylate ≥98.0% (by GC)
Ethyl-2-cyano-3,3-diphenylacrylate
Formula: C₁₈H₁₅NO₂
Peso molecolare: 277,32 g/mol
Punto di ebollizione: 200°C
MDL Number: MFCD00027364
CAS Number: 5232-99-5
EINECS: 226-029-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C
Solubility in Methanol within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR