Vinyl decanoate ≥99.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
Synonyms:
Decanoic Acid Vinyl Ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
C0895-25ML
C0895-500ML
TCIAC0895-25MLEA
24
EUR
TCIAC0895-25ML
TCIAC0895-500ML
Vinyl decanoate ≥99.0% (by GC) stabilizzato
Vinyl decanoate
Formula:
C₁₂H₂₂O₂ Peso molecolare: 198,31 g/mol Punto di ebollizione: 148°C Flash Pt: 106°C |
MDL Number:
MFCD00027349 CAS Number: 4704-31-8 EINECS: 225-185-7 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...