Phenyl-3-chloropropionate ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Phenyl 3-chloropropanoate, 3-Chloropropionic Acid Phenyl Ester
C0850-25ML
C0850-500ML
TCIAC0850-25MLEA
32
EUR
TCIAC0850-25ML
TCIAC0850-500ML
Phenyl-3-chloropropionate ≥95.0% (by GC)
Phenyl-3-chloropropionate
Formula:
C₉H₉ClO₂ Peso molecolare: 184,62 g/mol Punto di ebollizione: 157°C |
MDL Number:
MFCD00045298 CAS Number: 24552-27-0 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...