4-Cyano-1-cyclohexene ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
1,2,3,6-Tetrahydrobenzonitrile, 3-Cyclohexene-1-carbonitrile
C0837-25ML
C0837-5ML
TCIAC0837-25MLEA
197
EUR
TCIAC0837-25ML
TCIAC0837-5ML
4-Cyano-1-cyclohexene ≥98.0% (by GC)
4-Cyano-1-cyclohexene
Formula:
C₇H₉N Peso molecolare: 107,16 g/mol Punto di ebollizione: 60°C Flash Pt: 66°C |
MDL Number:
MFCD00013778 CAS Number: 100-45-8 UN: 3276 ADR: 6.1,III |
|
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...