Chelidamic acid monohydrate ≥95.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Oxo-1,4-dihydropyridine-2,6-dicarboxylic acid monohydrate
C0821-25G
C0821-5G
TCIAC0821-25GEA
172
EUR
TCIAC0821-25G
TCIAC0821-5G
Chelidamic acid monohydrate ≥95.0% (by titrimetric analysis)
Chelidamic acid monohydrate
Formula:
C₇H₅NO₅·H₂O Peso molecolare: 201,134 g/mol |
MDL Number:
MFCD00066478 CAS Number: 199926-39-1 EINECS: 205-335-8 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (Neutralization titration) | min. 95.0 % (calcd.on anh.substance) |
Water | max. 10.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...