Stampa…

β-Caryophyllene ≥90.0% (by GC)

Fornitore: TCI

Synonyms: (1R,4E,9S)-4,11,11-trimethyl-8-methylene-bicyclo[7.2.0]undec-4-ene, (1R,9S,E)-4,11,11-Trimethyl-8-methylenebicyclo[7.2.0]undec-4-ene, (−)-β-Caryophyllene, (-)-trans-caryophyllene, NSC 11906

C0796-25ML C0796-500ML C0796-100ML
TCIAC0796-25MLEA 23 EUR
TCIAC0796-25ML TCIAC0796-500ML TCIAC0796-100ML
β-Caryophyllene ≥90.0% (by GC)
β-Caryophyllene
Formula: C₁₅H₂₄
Peso molecolare: 204.36 g/mol
Punto di ebollizione: 116°C
Flash Pt: 97°C
MDL Number: MFCD00075925
CAS Number: 87-44-5
EINECS: 201-746-1
Merck Index: 14,01875

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %
Specific rotation [a]20/D -7.0 to -12.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR