1-Chlorodocosane ≥98.0% (by GC)
Fornitore: TCI
C0775-25G
C0775-5G
TCIAC0775-25GEA
148
EUR
TCIAC0775-25G
TCIAC0775-5G
1-Chlorodocosane ≥98.0% (by GC)
1-Chlorodocosane
Formula:
C₂₂H₄₅Cl |
MDL Number:
MFCD00039383 CAS Number: 42217-03-8 EINECS: 255-710-5 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 38.0 to 43.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...