Stampa…

Butyl-4-Carboxyphenyl carbonate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Carboxyphenyl Butyl Carbonate

C0773-1G C0773-5G
TCIAC0773-1GEA 97 EUR
TCIAC0773-1G TCIAC0773-5G
Butyl-4-Carboxyphenyl carbonate ≥98.0% (by titrimetric analysis)
Butyl-4-Carboxyphenyl carbonate
Formula: C₁₂H₁₄O₅
Peso molecolare: 238,24 g/mol
MDL Number: MFCD00060074
CAS Number: 14180-12-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 144.0 to 148.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR