N-Carbobenzyloxy-L-valine ≥99.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
N-Cbz-L-valine, Z-L-Val-OH, Z-Val-OH
C0761-25G
C0761-5G
TCIAC0761-25GEA
62
EUR
TCIAC0761-25G
TCIAC0761-5G
N-Carbobenzyloxy-L-valine ≥99.0% (by HPLC, titration analysis)
N-Carbobenzyloxy-L-valine
Formula:
C₁₃H₁₇NO₄ Peso molecolare: 251,3 g/mol |
MDL Number:
MFCD00008922 CAS Number: 1149-26-4 EINECS: 214-562-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 59.0 to 62.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...