Stampa…

2-Cinnamoylthiophene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Styryl 2-Thienyl Ketone, 3-Phenyl-1-(thiophen-2-yl)prop-2-en-1-one, 3-Phenyl-1-(2-thienyl)-2-propen-1-one

C0755-1G C0755-5G
TCIAC0755-1GEA 56 EUR
TCIAC0755-1G TCIAC0755-5G
2-Cinnamoylthiophene ≥98.0% (by GC)
2-Cinnamoylthiophene
Formula: C₁₃H₁₀OS
Peso molecolare: 214,28 g/mol
MDL Number: MFCD00059742
CAS Number: 3988-77-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point gt. 81.0 to 85.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR