N-Carbobenzoxy-4-aminobutyric acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
N-Carbobenzoxy-4-aminobutyric acid, N-Cbz-4-aminobutanoic acid, 4-(Benzyloxycarbonylamino)butyric acid, Z-GABA-OH, N-Cbz-4-aminobutyric acid, Z-γ-Abu-OH
C0753-1G
C0753-25G
TCIAC0753-1GEA
16
EUR
TCIAC0753-1G
TCIAC0753-25G
N-Carbobenzoxy-4-aminobutyric acid ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-4-aminobutyric acid
Formula:
C₁₂H₁₅NO₄ Peso molecolare: 237,26 g/mol |
MDL Number:
MFCD00055835 CAS Number: 5105-78-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 63.0 to 68.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...