N-Carbobenzoxy-D-methionine ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
N-Benzyloxycarbonyl-D-methionine, Cbz-D-Met-OH, Z-D-Met-OH, N-Cbz-D-methionine
C0665-100MG
C0665-25G
C0665-5G
TCIAC0665-100MGEA
16
EUR
TCIAC0665-100MG
TCIAC0665-25G
TCIAC0665-5G
N-Carbobenzoxy-D-methionine ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-D-methionine
Formula:
C₁₃H₁₇NO₄S Peso molecolare: 283,34 g/mol |
MDL Number:
MFCD00026043 CAS Number: 28862-80-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Specific rotation [a]20/D | +24.0 to +28.0 deg (C=1, MeOH) |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...