N-Carbobenzyloxy-D-alanine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
Z-D-Ala-OH, N-Cbz-D-alanine
C0664-1G
C0664-25G
C0664-5G
TCIAC0664-1GEA
16
EUR
TCIAC0664-1G
TCIAC0664-25G
TCIAC0664-5G
N-Carbobenzyloxy-D-alanine ≥98.0% (by HPLC, titration analysis)
N-Carbobenzyloxy-D-alanine
Formula:
C₁₁H₁₃NO₄ Peso molecolare: 223,2 g/mol |
MDL Number:
MFCD00063126 CAS Number: 26607-51-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | +10.0 to +20.0 deg (C=2, AcOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...