Stampa…

(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Cbz-Phe-OH, N-Cbz-L-phenylalanine, N-Carbobenzyloxy-L-phenylalanine, Z-Phe-OH

C0660-25G C0660-5G
TCIAC0660-25GEA 64 EUR
TCIAC0660-25G TCIAC0660-5G
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid ≥98.0% (by HPLC, titration analysis)
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid
Formula: C₁₇H₁₇NO₄
Peso molecolare: 299,3 g/mol
MDL Number: MFCD00020418
CAS Number: 1161-13-3
EINECS: 214-599-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR