Stampa…

N-Cbz-L-serine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: N-Carbobenzyloxy-L-serine, Z-Ser-OH, N-Cbz-L-serine

C0635-25G C0635-5G
TCIAC0635-25GEA 54 EUR
TCIAC0635-25G TCIAC0635-5G
N-Cbz-L-serine ≥98.0% (by HPLC, titration analysis)
N-Cbz-L-serine
Formula: C₁₁H₁₃NO₅
Peso molecolare: 239,2 g/mol
MDL Number: MFCD00002662
CAS Number: 1145-80-8
EINECS: 214-546-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 117.0 to 120.0 deg-C
Specific rotation [a]20/D +5.6 to +6.6 deg (C=6, AcOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR