Stampa…

Methyl-4-chloro-2-methylphenoxyacetate ≥94.0% (by GC)

Fornitore: TCI

C0598-25G C0598-1G
TCIAC0598-25GEA 192 EUR
TCIAC0598-25G TCIAC0598-1G
Methyl-4-chloro-2-methylphenoxyacetate ≥94.0% (by GC)
Methyl-4-chloro-2-methylphenoxyacetate
Formula: C₁₀H₁₁ClO₃
Punto di ebollizione: 127°C
MDL Number: MFCD00055259
CAS Number: 2436-73-9
EINECS: 219-430-7

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR