Stampa…

L-Cysteine methyl ester hydrochloride ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: H-Cys-OMe·HCl, (R)-Methyl 2-amino-3-mercaptopropanoate hydrochloride

C0577-5G C0577-25G C0577-500G
TCIAC0577-5GEA 16 EUR
TCIAC0577-5G TCIAC0577-25G TCIAC0577-500G
L-Cysteine methyl ester hydrochloride ≥99.0% (by titrimetric analysis)
L-Cysteine methyl ester hydrochloride
Formula: C₄H₉NO₂S·HCl
Peso molecolare: 171,7 g/mol
MDL Number: MFCD00038998
CAS Number: 18598-63-5
EINECS: 242-435-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (Iodometric Titration) min. 99.0 %
Specific rotation [a]20/D -1.5 to -3.5 deg (C=20, MeOH)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR