Stampa…

N-Carbobenzoxy-L-glutamine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: N-Cbz-L-glutamine, Z-Gln-OH

C0574-25G C0574-5G
TCIAC0574-25GEA 107 EUR
TCIAC0574-25G TCIAC0574-5G
N-Carbobenzoxy-L-glutamine ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-L-glutamine
Formula: C₁₃H₁₆N₂O₅
Peso molecolare: 280,28 g/mol
MDL Number: MFCD00008043
CAS Number: 2650-64-8
EINECS: 220-173-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C
Specific rotation [a]20/D -6.5 to -8.5 deg (C=2, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR