Chrome yellow ≥95.0% (by HPLC)
Fornitore: TCI
Formula:
C₁₇H₁₂N₂O₆S |
MDL Number:
MFCD00021624 CAS Number: 6054-97-3 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Lambda max. | 358.0 to 362.0 nm (H2O) |
Absorbance (E1%1cm) | min. 350 (H2O, 358.0 to 362.0 nm) |
Drying loss | max. 8.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...