Stampa…

Isopropyl cyclohexanecarboxylate ≥98.0% (by GC)

Fornitore: TCI

Synonyms: Cyclohexanecarboxylic Acid Isopropyl Ester

C0472-25ML C0472-500ML
TCIAC0472-25MLEA 41 EUR
TCIAC0472-25ML TCIAC0472-500ML
Isopropyl cyclohexanecarboxylate ≥98.0% (by GC)
Isopropyl cyclohexanecarboxylate
Formula: C₁₀H₁₈O₂
Peso molecolare: 170.25 g/mol
Punto di ebollizione: 131°C
MDL Number: MFCD00058949
CAS Number: 6553-80-6
EINECS: 229-474-9

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR