Stampa…

Crotonoyl chloride cis- and trans-mixture ≥95.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: But-2-enoyl chloride, 2-Butenoyl chloride, Crotonoyl Chloride, Crotonic acid chloride

C0424-25G C0424-500G C0424-100G
TCIAC0424-25GEA 32 EUR
TCIAC0424-25G TCIAC0424-500G TCIAC0424-100G
Crotonoyl chloride cis- and trans-mixture ≥95.0% (by GC)
Crotonoyl chloride
Formula: C₄H₅ClO
Peso molecolare: 104,53 g/mol
Punto di ebollizione: 125°C
Flash Pt: 35°C
MDL Number: MFCD00000734
CAS Number: 10487-71-5
EINECS: 234-010-3
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8339966514f43cacc60735c7282228908eb24ad8&vid=d829646289c5a5ef5ee441ac38c1064fe11fd300&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 % (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR